Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACTAATTTGTGGATTGGATATT[G/T]TGCCTTTCACCTGGGTGACTACAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TTC26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TTC26 - tetratricopeptide repeat domain 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144920.2 | 240 | Missense Mutation | TGT,TTT | C,F 66 | NP_001138392.1 | |
NM_001144923.2 | 240 | Intron | NP_001138395.1 | |||
NM_001287512.1 | 240 | Missense Mutation | TGT,TTT | C,F 66 | NP_001274441.1 | |
NM_001287513.1 | 240 | UTR 5 | NP_001274442.1 | |||
NM_001318333.1 | 240 | UTR 5 | NP_001305262.1 | |||
NM_001321740.1 | 240 | Missense Mutation | TGT,TTT | C,F 66 | NP_001308669.1 | |
NM_001321741.1 | 240 | Missense Mutation | TGT,TTT | C,F 66 | NP_001308670.1 | |
NM_001321742.1 | 240 | Missense Mutation | TGT,TTT | C,F 66 | NP_001308671.1 | |
NM_024926.3 | 240 | Missense Mutation | TGT,TTT | C,F 66 | NP_079202.2 |