Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTCGGAGAAGGACAGGACAATG[A/G]CGGCCTTAGGGTCCCCGTCGCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf55-LUC7L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf55-LUC7L2 - C7orf55-LUC7L2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244584.2 | 257 | Missense Mutation | ACG,GCG | T,A 2 | NP_001231513.1 |
FMC1 - formation of mitochondrial complex V assembly factor 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_197964.4 | 257 | Missense Mutation | ACG,GCG | T,A 2 | NP_932068.2 |
LUC7L2 - LUC7 like 2, pre-mRNA splicing factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244585.1 | 257 | Intron | NP_001231514.1 | |||
NM_001270643.1 | 257 | Intron | NP_001257572.1 | |||
NM_016019.4 | 257 | Intron | NP_057103.2 |