Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGAATGTAGCACAGAGAGATG[C/T]GGCATCCCATTAGTGACAGTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615137 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WBSCR17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WBSCR17 - Williams-Beuren syndrome chromosome region 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022479.2 | Intron | NP_071924.1 | ||||
XM_011516467.2 | Intron | XP_011514769.1 | ||||
XM_011516469.2 | Intron | XP_011514771.1 | ||||
XM_017012521.1 | Intron | XP_016868010.1 |