Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAGAGATGGGTGAGTCTATGCC[C/T]GGCATGGGGGTTGTGCTCAACTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALN1 - calneuron 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017440.2 | 6966 | UTR 3 | NP_001017440.1 | |||
NM_031468.3 | 6966 | UTR 3 | NP_113656.2 | |||
XM_011516594.2 | 6966 | UTR 3 | XP_011514896.1 | |||
XM_011516595.1 | 6966 | UTR 3 | XP_011514897.1 | |||
XM_011516596.2 | 6966 | UTR 3 | XP_011514898.1 | |||
XM_011516597.2 | 6966 | UTR 3 | XP_011514899.1 | |||
XM_011516599.1 | 6966 | UTR 3 | XP_011514901.1 | |||
XM_017012675.1 | 6966 | UTR 3 | XP_016868164.1 | |||
XM_017012676.1 | 6966 | UTR 3 | XP_016868165.1 | |||
XM_017012677.1 | 6966 | UTR 3 | XP_016868166.1 | |||
XM_017012678.1 | 6966 | UTR 3 | XP_016868167.1 | |||
XM_017012679.1 | 6966 | UTR 3 | XP_016868168.1 | |||
XM_017012680.1 | 6966 | UTR 3 | XP_016868169.1 | |||
XM_017012681.1 | 6966 | UTR 3 | XP_016868170.1 | |||
XM_017012682.1 | 6966 | UTR 3 | XP_016868171.1 | |||
XM_017012683.1 | 6966 | UTR 3 | XP_016868172.1 |