Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCATCATCTTGCAGAAATCCTGT[C/T]CCACATGACTTCTCTCCTTCATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO43 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO43 - F-box protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029860.3 | 1584 | Intron | NP_001025031.2 | |||
XM_011516987.2 | 1584 | Silent Mutation | GGA,GGG | G,G 544 | XP_011515289.1 | |
XM_011516989.2 | 1584 | Silent Mutation | GGA,GGG | G,G 510 | XP_011515291.1 | |
XM_011516990.1 | 1584 | Silent Mutation | GGA,GGG | G,G 510 | XP_011515292.1 | |
XM_011516992.2 | 1584 | Silent Mutation | GGA,GGG | G,G 544 | XP_011515294.1 | |
XM_017013358.1 | 1584 | Silent Mutation | GGA,GGG | G,G 544 | XP_016868847.1 |