Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACGGAGTGACTCTGGAGACTCG[A/G]GCATATGAAGAAGTTCTGAATTATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604679 | ||||||||||||||||||||
Literature Links: |
MIR7705 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR7705 - microRNA 7705 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PABPC1 - poly(A) binding protein cytoplasmic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002568.3 | 3205 | Intron | NP_002559.2 | |||
XM_005250861.2 | 3205 | Missense Mutation | CCC,CTC | P,L 597 | XP_005250918.1 |