Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAATTTGTCCTTGTGTGGTAAGA[C/T]TGTTCCATCAATGGCCCCCAATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606109 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDCD1 - NudC domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128211.1 | 2692 | UTR 3 | NP_001121683.1 | |||
NM_032869.3 | 2692 | UTR 3 | NP_116258.2 | |||
XM_017013910.1 | 2692 | UTR 3 | XP_016869399.1 | |||
XM_017013911.1 | 2692 | UTR 3 | XP_016869400.1 |