Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACCCTAAGCACAAACTAGTACA[C/T]TCTTTAATTTTTATTGCAGGACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120324 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL14A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL14A1 - collagen type XIV alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021110.3 | Intron | NP_066933.1 | ||||
XM_005251059.3 | Intron | XP_005251116.1 | ||||
XM_006716651.3 | Intron | XP_006716714.1 | ||||
XM_017013809.1 | Intron | XP_016869298.1 |