Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACATGGCAACAAAATGCACCCA[A/G]TTTACATGCATCAACAAGTTAATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616609 | ||||||||||||||||||||
Literature Links: |
TMEM65 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM65 - transmembrane protein 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194291.2 | 3751 | UTR 3 | NP_919267.2 | |||
XM_011516847.2 | 3751 | Intron | XP_011515149.1 | |||
XM_017013104.1 | 3751 | Intron | XP_016868593.1 | |||
XM_017013105.1 | 3751 | Intron | XP_016868594.1 |