Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAAGAATGTCAAGTCTTTGAAG[G/T]TACACTAAATTTTTTTTTTTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609864 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf88 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf88 - chromosome 8 open reading frame 88 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM55A - transmembrane protein 55A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018710.2 | 1517 | UTR 3 | NP_061180.1 | |||
XM_011517152.2 | 1517 | Intron | XP_011515454.1 |