Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGCCTGTTCTCTTTGTCAACAC[A/G]TTAAATAAAGGAAGAAAAATAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC69 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC69 - leucine rich repeat containing 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129890.1 | 1194 | UTR 3 | NP_001123362.1 |
SLC26A7 - solute carrier family 26 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282356.1 | 1194 | Intron | NP_001269285.1 | |||
NM_001282357.1 | 1194 | Intron | NP_001269286.1 | |||
NM_052832.3 | 1194 | Intron | NP_439897.1 | |||
NM_134266.1 | 1194 | Intron | NP_599028.1 |