Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCAGCTGTCGGTGACCCTGTCC[A/G]CGGTGTCTGTGTTCACTTTTGTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138251 MIM: 609564 | ||||||||||||||||||||
Literature Links: |
GRINA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRINA - glutamate ionotropic receptor NMDA type subunit associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000837.1 | 701 | Missense Mutation | ACG,GCG | T,A 179 | NP_000828.1 | |
NM_001009184.1 | 701 | Missense Mutation | ACG,GCG | T,A 179 | NP_001009184.1 |
PARP10 - poly(ADP-ribose) polymerase family member 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |