Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAATCTCTCTTCAGTGCCTTCC[A/G]CTAGTACTACTTGTAAAAGAAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM92A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM92A1 - family with sequence similarity 92 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283034.1 | Intron | NP_001269963.1 | ||||
NM_145269.4 | Intron | NP_660312.2 | ||||
XM_005250787.2 | Intron | XP_005250844.1 |
LINC00535 - long intergenic non-protein coding RNA 535 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |