Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTGTCTAGTGTTTTGGGTTCC[C/T]TACACCAGGATTGTGGAGGAAGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604207 MIM: 611871 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPNE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPNE3 - copine 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003909.4 | Intron | NP_003900.1 | ||||
XM_005251093.4 | Intron | XP_005251150.1 | ||||
XM_017013945.1 | Intron | XP_016869434.1 |