Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTGAAGACTTAAGAGATGAGAA[C/T]CAGGTGTCTTTTATTGCTGTGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBXN2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBXN2B - UBX domain protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077619.1 | Intron | NP_001071087.1 | ||||
XM_005251165.2 | Intron | XP_005251222.1 | ||||
XM_005251166.1 | Intron | XP_005251223.1 |