Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCTACTTAAGCTGGTTGTGTTT[C/T]ACAGGGAACACTGTTTTCCTAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C8orf22 - chromosome 8 open reading frame 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007176.4 | Intron | NP_001007177.1 | ||||
NM_001256596.1 | Intron | NP_001243525.1 | ||||
NM_001256597.1 | Intron | NP_001243526.1 | ||||
NM_001256598.1 | Intron | NP_001243527.1 |