Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAATCCAGTGATTCCCAGATTTA[C/T]TTGGTTATGACCATCTATCCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAM1 - translocation associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317804.1 | 3070 | UTR 3 | NP_001304733.1 | |||
NM_001317805.1 | 3070 | UTR 3 | NP_001304734.1 | |||
NM_014294.5 | 3070 | UTR 3 | NP_055109.1 | |||
XM_017013266.1 | 3070 | Intron | XP_016868755.1 | |||
XM_017013267.1 | 3070 | Intron | XP_016868756.1 |