Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTGAACTATTATTGTAACTCA[A/G]CAATCAAGAGTCCTAGCTTTTAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606598 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDAP1 - ganglioside induced differentiation associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040875.2 | Intron | NP_001035808.1 | ||||
NM_018972.2 | Intron | NP_061845.2 | ||||
XM_011517551.2 | Intron | XP_011515853.1 | ||||
XM_011517552.2 | Intron | XP_011515854.1 | ||||
XM_017013585.1 | Intron | XP_016869074.1 | ||||
XM_017013586.1 | Intron | XP_016869075.1 |