Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGCATTTATATTAATCAGAGAA[C/G]TAGATAATGTCATTCTTTTTGTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL7 - interleukin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000880.3 | Intron | NP_000871.1 | ||||
NM_001199886.1 | Intron | NP_001186815.1 | ||||
NM_001199887.1 | Intron | NP_001186816.1 | ||||
NM_001199888.1 | Intron | NP_001186817.1 | ||||
XM_011517522.2 | Intron | XP_011515824.1 | ||||
XM_011517523.2 | Intron | XP_011515825.1 | ||||
XM_017013397.1 | Intron | XP_016868886.1 |
ZC2HC1A - zinc finger C2HC-type containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016010.2 | Intron | NP_057094.2 | ||||
XM_005251256.1 | Intron | XP_005251313.1 | ||||
XM_006716454.2 | Intron | XP_006716517.1 | ||||
XM_011517539.2 | Intron | XP_011515841.1 |