Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTACAATTAAAAAAGAACAA[A/G]GACAATTTGATAAGTGCCTTTAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAG1 - phosphoprotein membrane anchor with glycosphingolipid microdomains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018440.3 | 10656 | UTR 3 | NP_060910.3 | |||
XM_017013640.1 | 10656 | UTR 3 | XP_016869129.1 | |||
XM_017013641.1 | 10656 | UTR 3 | XP_016869130.1 | |||
XM_017013642.1 | 10656 | UTR 3 | XP_016869131.1 | |||
XM_017013643.1 | 10656 | UTR 3 | XP_016869132.1 |