Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCAGTTGTTTCATTTTCTTAAA[C/T]ATTTCAAACATTAAAACAAAAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1H - ATPase H+ transporting V1 subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015941.3 | Intron | NP_057025.2 | ||||
NM_213619.2 | Intron | NP_998784.1 | ||||
NM_213620.2 | Intron | NP_998785.1 | ||||
XM_006716455.2 | Intron | XP_006716518.1 | ||||
XM_011517542.1 | Intron | XP_011515844.1 |