Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATTTTCACATTGTTTAATTTTA[C/T]GTACATTTCACATATCATAAATACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM68 - transmembrane protein 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286657.1 | 2590 | UTR 3 | NP_001273586.1 | |||
NM_001286660.1 | 2590 | Intron | NP_001273589.1 | |||
NM_001286661.1 | 2590 | UTR 3 | NP_001273590.1 | |||
NM_152417.2 | 2590 | UTR 3 | NP_689630.1 | |||
XM_005251150.1 | 2590 | UTR 3 | XP_005251207.1 | |||
XM_011517454.1 | 2590 | UTR 3 | XP_011515756.1 | |||
XM_011517455.2 | 2590 | Intron | XP_011515757.1 | |||
XM_017013040.1 | 2590 | UTR 3 | XP_016868529.1 |