Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCGGCGGCTGCAGTAGCAGCAG[C/T]AGCTACTGCAGCTTCCCTCCCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LURAP1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LURAP1L - leucine rich adaptor protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203403.1 | 883 | Silent Mutation | AGC,AGT | S,S 60 | NP_981948.1 | |
XM_005251443.2 | 883 | Silent Mutation | AGC,AGT | S,S 60 | XP_005251500.1 |
LURAP1L-AS1 - LURAP1L antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |