Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTTGGCTTCTACTTTCTACCC[C/T]TGCACATTGCTCTGGCCTGGTGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMARCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMARCA2 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289396.1 | Intron | NP_001276325.1 | ||||
NM_001289397.1 | Intron | NP_001276326.1 | ||||
NM_001289398.1 | Intron | NP_001276327.1 | ||||
NM_001289399.1 | Intron | NP_001276328.1 | ||||
NM_001289400.1 | Intron | NP_001276329.1 | ||||
NM_003070.4 | Intron | NP_003061.3 | ||||
NM_139045.3 | Intron | NP_620614.2 |