Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTTTCTGTGGGCTATGAGATCA[C/T]AGTTAGCTAGTCTGCATTACTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613747 MIM: 602852 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF24 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF24 - kinesin family member 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT2 - nudix hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161.4 | Intron | NP_001152.1 | ||||
NM_001244390.1 | Intron | NP_001231319.1 | ||||
NM_147172.2 | Intron | NP_671701.1 | ||||
NM_147173.2 | Intron | NP_671702.1 |