Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATCTCATAATAGGAAAGGCCTTC[A/G]GCATAAAAGCAGATACAGAAATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLTA - clathrin light chain A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076677.2 | Intron | NP_001070145.1 | ||||
NM_001184760.1 | Intron | NP_001171689.1 | ||||
NM_001184761.1 | Intron | NP_001171690.1 | ||||
NM_001184762.1 | Intron | NP_001171691.1 | ||||
NM_001311203.1 | Intron | NP_001298132.1 | ||||
NM_001311204.1 | Intron | NP_001298133.1 | ||||
NM_001311205.1 | Intron | NP_001298134.1 | ||||
NM_001311206.1 | Intron | NP_001298135.1 | ||||
NM_001833.3 | Intron | NP_001824.1 | ||||
NM_007096.3 | Intron | NP_009027.1 | ||||
XM_017014257.1 | Intron | XP_016869746.1 | ||||
XM_017014258.1 | Intron | XP_016869747.1 |