Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGAGGTTTTAATCCATGAACAT[G/T]ATCTGTCTCTCTACTTACTTAAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCL1 - RNA terminal phosphate cyclase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286699.1 | Intron | NP_001273628.1 | ||||
NM_001286700.1 | Intron | NP_001273629.1 | ||||
NM_001286701.1 | Intron | NP_001273630.1 | ||||
NM_005772.4 | Intron | NP_005763.3 | ||||
XM_006716715.3 | Intron | XP_006716778.1 | ||||
XM_011517673.2 | Intron | XP_011515975.1 | ||||
XM_017014176.1 | Intron | XP_016869665.1 |