Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCCCTTTCCCATTTATAGAGA[A/G]AAAAGAATCCTGAGGCAAACTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608678 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL33 - interleukin 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199640.1 | Intron | NP_001186569.1 | ||||
NM_001199641.1 | Intron | NP_001186570.1 | ||||
NM_001314044.1 | Intron | NP_001300973.1 | ||||
NM_001314045.1 | Intron | NP_001300974.1 | ||||
NM_001314046.1 | Intron | NP_001300975.1 | ||||
NM_001314047.1 | Intron | NP_001300976.1 | ||||
NM_001314048.1 | Intron | NP_001300977.1 | ||||
NM_033439.3 | Intron | NP_254274.1 | ||||
XM_011518061.2 | Intron | XP_011516363.1 | ||||
XM_017015285.1 | Intron | XP_016870774.1 |