Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCGCTCCCGCCCTCCGAGGTG[A/G]GGCTAGAGGGGAGGGTTCGACTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602424 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMRT1 - doublesex and mab-3 related transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021951.2 | Intron | NP_068770.2 | ||||
XM_006716732.1 | Intron | XP_006716795.1 | ||||
XM_011517770.1 | Intron | XP_011516072.1 | ||||
XM_011517771.1 | Intron | XP_011516073.1 | ||||
XM_011517772.1 | Intron | XP_011516074.1 | ||||
XM_011517773.2 | Intron | XP_011516075.1 | ||||
XM_017014374.1 | Intron | XP_016869863.1 | ||||
XM_017014375.1 | Intron | XP_016869864.1 |