Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGCCAAGGATGTTTGGGATTTA[C/G]CCATTGCAACAATTCTTCATCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASEF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASEF - RAS and EF-hand domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152573.3 | 2461 | Silent Mutation | GGC,GGG | G,G 740 | NP_689786.2 | |
XM_005251730.3 | 2461 | Silent Mutation | GGC,GGG | G,G 717 | XP_005251787.1 | |
XM_005251731.3 | 2461 | Silent Mutation | GGC,GGG | G,G 611 | XP_005251788.1 |