Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGGATAAGGTGGCATGGACCC[G/T]CCATGGCGCGGCTCTGGGGCTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600456 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTRK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NTRK2 - neurotrophic receptor tyrosine kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007097.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | NP_001007098.1 | |
NM_001018064.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | NP_001018074.1 | |
NM_001018065.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | NP_001018075.1 | |
NM_001018066.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | NP_001018076.1 | |
NM_001291937.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | NP_001278866.1 | |
NM_006180.4 | 943 | Missense Mutation | GCC,TCC | A,S 11 | NP_006171.2 | |
XM_005252001.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_005252058.1 | |
XM_005252003.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_005252060.1 | |
XM_005252004.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_005252061.1 | |
XM_005252006.3 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_005252063.1 | |
XM_005252007.3 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_005252064.1 | |
XM_011518718.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_011517020.1 | |
XM_011518720.2 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_011517022.1 | |
XM_017014751.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870240.1 | |
XM_017014752.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870241.1 | |
XM_017014753.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870242.1 | |
XM_017014754.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870243.1 | |
XM_017014755.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870244.1 | |
XM_017014756.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870245.1 | |
XM_017014757.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870246.1 | |
XM_017014758.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870247.1 | |
XM_017014759.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870248.1 | |
XM_017014760.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870249.1 | |
XM_017014761.1 | 943 | Missense Mutation | GCC,TCC | A,S 11 | XP_016870250.1 |