Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGGAAGGTTTGCTTCTCCCCTC[G/C]GAAGAGCTAATTTTGAAGATGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR13C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR13C2 - olfactory receptor family 13 subfamily C member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR13C5 - olfactory receptor family 13 subfamily C member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004482.1 | 693 | Silent Mutation | TCC,TCG | S,S 231 | NP_001004482.1 |