Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTCTTGGCAGTGTCTGGGTAT[A/C]ATGATTTTCCATCAAAATAATTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC44A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC44A1 - solute carrier family 44 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286730.1 | Intron | NP_001273659.1 | ||||
NM_080546.4 | Intron | NP_536856.2 | ||||
XM_005251855.4 | Intron | XP_005251912.1 | ||||
XM_006717027.3 | Intron | XP_006717090.1 | ||||
XM_006717028.3 | Intron | XP_006717091.1 | ||||
XM_006717029.2 | Intron | XP_006717092.1 | ||||
XM_017014559.1 | Intron | XP_016870048.1 | ||||
XM_017014560.1 | Intron | XP_016870049.1 |