Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAACGGGAGTGGCTGCTCAGGGG[A/G]ACATTCTGGGGCTCTTCCCTCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607440 MIM: 186855 | ||||||||||||||||||||
Literature Links: |
FKTN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FKTN - fukutin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAL2 - TAL bHLH transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005421.2 | 257 | Missense Mutation | AAC,GAC | N,D 73 | NP_005412.1 |