Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTGGGTGGAGTCTCACTGGAGT[G/T]TTTTTTTTAGGGTGTCTGAAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GDA - guanine deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242505.2 | Intron | NP_001229434.1 | ||||
NM_001242506.2 | Intron | NP_001229435.1 | ||||
NM_001242507.2 | Intron | NP_001229436.1 | ||||
NM_004293.4 | Intron | NP_004284.1 | ||||
XM_005252317.1 | Intron | XP_005252374.1 | ||||
XM_011519213.1 | Intron | XP_011517515.1 | ||||
XM_011519214.1 | Intron | XP_011517516.1 | ||||
XM_011519215.1 | Intron | XP_011517517.1 | ||||
XM_011519216.1 | Intron | XP_011517518.1 | ||||
XM_011519217.1 | Intron | XP_011517519.1 | ||||
XM_011519218.2 | Intron | XP_011517520.1 | ||||
XM_011519219.1 | Intron | XP_011517521.1 | ||||
XM_011519220.1 | Intron | XP_011517522.1 | ||||
XM_011519221.1 | Intron | XP_011517523.1 | ||||
XM_011519222.1 | Intron | XP_011517524.1 | ||||
XM_011519223.2 | Intron | XP_011517525.1 | ||||
XM_017015333.1 | Intron | XP_016870822.1 | ||||
XM_017015334.1 | Intron | XP_016870823.1 | ||||
XM_017015335.1 | Intron | XP_016870824.1 | ||||
XM_017015336.1 | Intron | XP_016870825.1 | ||||
XM_017015337.1 | Intron | XP_016870826.1 | ||||
XM_017015338.1 | Intron | XP_016870827.1 | ||||
XM_017015339.1 | Intron | XP_016870828.1 | ||||
XM_017015340.1 | Intron | XP_016870829.1 |