Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTTTGTGAAGATGAACAGGACC[A/G]TGCTTGTAAAGGGCTTAGAACAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL27A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL27A1 - collagen type XXVII alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032888.3 | Intron | NP_116277.2 | ||||
XM_006717308.2 | Intron | XP_006717371.1 | ||||
XM_006717310.3 | Intron | XP_006717373.1 | ||||
XM_011519138.2 | Intron | XP_011517440.1 | ||||
XM_011519140.1 | Intron | XP_011517442.1 | ||||
XM_011519142.2 | Intron | XP_011517444.1 | ||||
XM_011519143.2 | Intron | XP_011517445.1 | ||||
XM_011519144.2 | Intron | XP_011517446.1 | ||||
XM_011519145.2 | Intron | XP_011517447.1 | ||||
XM_017015239.1 | Intron | XP_016870728.1 |