Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCATCATTGACATTCTAACTAAG[C/T]GAAACAATGCACAGCGTCAACAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA1 - annexin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000700.2 | 368 | Nonsense Mutation | CGA,TGA | R,* 72 | NP_000691.1 | |
XM_011518609.1 | 368 | Nonsense Mutation | CGA,TGA | R,* 72 | XP_011516911.1 | |
XM_017014657.1 | 368 | Nonsense Mutation | CGA,TGA | R,* 83 | XP_016870146.1 |