Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCGCGGCCGAACTCACCTCGTGG[C/T]CTCGGCGTGGTGCTCTCAGCTCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604602 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRRF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRRF - mitochondrial ribosome recycling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173512.1 | 120 | UTR 5 | NP_001166983.1 | |||
NM_138777.3 | 120 | UTR 5 | NP_620132.1 | |||
NM_199177.2 | 120 | Intron | NP_954646.1 | |||
XM_005252302.3 | 120 | Intron | XP_005252359.1 | |||
XM_011519183.2 | 120 | Intron | XP_011517485.1 | |||
XM_011519184.2 | 120 | Intron | XP_011517486.1 | |||
XM_011519185.2 | 120 | Intron | XP_011517487.1 | |||
XM_011519186.1 | 120 | Intron | XP_011517488.1 | |||
XM_011519187.2 | 120 | UTR 5 | XP_011517489.1 | |||
XM_017015286.1 | 120 | Intron | XP_016870775.1 | |||
XM_017015287.1 | 120 | Intron | XP_016870776.1 |
RBM18 - RNA binding motif protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033117.3 | 120 | UTR 5 | NP_149108.1 | |||
XM_006717319.1 | 120 | UTR 5 | XP_006717382.1 |