Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGGTCTTCAGATTGAGAGGAAC[A/C]GTACACAGGGAACTGCATCTGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR1L8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR1L8 - olfactory receptor family 1 subfamily L member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004454.1 | 1932 | Intron | NP_001004454.1 | |||
XM_017014284.1 | 1932 | Intron | XP_016869773.1 | |||
XM_017014285.1 | 1932 | UTR 3 | XP_016869774.1 | |||
XM_017014286.1 | 1932 | Intron | XP_016869775.1 |
OR1N2 - olfactory receptor family 1 subfamily N member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |