Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGACACTGCTGGTGTGGTTGATT[A/C]TTTCCATTGACTTGGGCTCAGTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR1B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR1B1 - olfactory receptor family 1 subfamily B member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR1L8 - olfactory receptor family 1 subfamily L member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004454.1 | 415 | Missense Mutation | NP_001004454.1 | |||
XM_017014284.1 | 415 | Missense Mutation | XP_016869773.1 | |||
XM_017014285.1 | 415 | Missense Mutation | XP_016869774.1 | |||
XM_017014286.1 | 415 | Missense Mutation | XP_016869775.1 |