Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTCATTTCTCCGTGAACCCGG[A/G]ATGACAACCTTGGCTAGGTGCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MVB12B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MVB12B - multivesicular body subunit 12B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011703.2 | Intron | NP_001011703.1 | ||||
NM_033446.2 | Intron | NP_258257.1 | ||||
XM_005252297.1 | Intron | XP_005252354.1 | ||||
XM_017015275.1 | Intron | XP_016870764.1 | ||||
XM_017015276.1 | Intron | XP_016870765.1 | ||||
XM_017015277.1 | Intron | XP_016870766.1 | ||||
XM_017015278.1 | Intron | XP_016870767.1 | ||||
XM_017015279.1 | Intron | XP_016870768.1 |