Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCAGCCAACAGAGACCTGGTCA[G/T]GTGCAGTGGCGGGTGGCTGAGGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610933 MIM: 180475 MIM: 194552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRSAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRSAM1 - leucine rich repeat and sterile alpha motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL12 - ribosomal protein L12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000976.3 | Intron | NP_000967.1 |
SNORA65 - small nucleolar RNA, H/ACA box 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF79 - zinc finger protein 79 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |