Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGGAAACCGAGACCAGCTAGAC[A/G]GTGGCAAAGCAGTGATTCAGACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600488 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCSK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCSK5 - proprotein convertase subtilisin/kexin type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190482.1 | Intron | NP_001177411.1 | ||||
NM_006200.5 | Intron | NP_006191.2 | ||||
XM_005252039.3 | Intron | XP_005252096.1 | ||||
XM_011518769.2 | Intron | XP_011517071.1 | ||||
XM_011518770.2 | Intron | XP_011517072.1 | ||||
XM_017014800.1 | Intron | XP_016870289.1 |