Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGTTTGCACAGATCCTAGATCCC[C/T]GTATATAATGGTATGCCTGTGATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611025 MIM: 611260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENKUR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENKUR - enkurin, TRPC channel interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270383.1 | Intron | NP_001257312.1 | ||||
NM_145010.3 | Intron | NP_659447.1 | ||||
XM_006717389.1 | Intron | XP_006717452.1 | ||||
XM_006717390.1 | Intron | XP_006717453.1 | ||||
XM_011519348.1 | Intron | XP_011517650.1 | ||||
XM_017015830.1 | Intron | XP_016871319.1 |
THNSL1 - threonine synthase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024838.4 | Intron | NP_079114.3 | ||||
XM_005252597.2 | Intron | XP_005252654.1 | ||||
XM_017016665.1 | Intron | XP_016872154.1 |