Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTTTAATGAGACTGCAAACCCG[C/T]CCACCCCCATAGTCTTCCCTAGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKR1E2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKR1E2 - aldo-keto reductase family 1 member E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040177.2 | Intron | NP_001035267.1 | ||||
NM_001271021.1 | Intron | NP_001257950.1 | ||||
NM_001271025.1 | Intron | NP_001257954.1 | ||||
XM_011519715.2 | Intron | XP_011518017.1 | ||||
XM_011519718.2 | Intron | XP_011518020.1 | ||||
XM_011519719.2 | Intron | XP_011518021.1 | ||||
XM_011519720.2 | Intron | XP_011518022.1 | ||||
XM_011519722.2 | Intron | XP_011518024.1 | ||||
XM_011519724.2 | Intron | XP_011518026.1 | ||||
XM_011519725.2 | Intron | XP_011518027.1 | ||||
XM_011519729.2 | Intron | XP_011518031.1 | ||||
XM_017016743.1 | Intron | XP_016872232.1 | ||||
XM_017016744.1 | Intron | XP_016872233.1 | ||||
XM_017016745.1 | Intron | XP_016872234.1 |