Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGTAGATATCTAGAAACAAATGA[C/T]ACCAGCTCACTGCCTCAAGTGTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM208B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM208B - family with sequence similarity 208 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321783.1 | Intron | NP_001308712.1 | ||||
NM_001321784.1 | Intron | NP_001308713.1 | ||||
NM_001321785.1 | Intron | NP_001308714.1 | ||||
NM_017782.4 | Intron | NP_060252.4 |
GDI2 - GDP dissociation inhibitor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115156.1 | Intron | NP_001108628.1 | ||||
NM_001494.3 | Intron | NP_001485.2 | ||||
XM_017016071.1 | Intron | XP_016871560.1 |