Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACTGGTGCCGTAGGCATGGCTT[G/T]TGCTATTAGTATCTTACTGAAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 150000 MIM: 150150 | ||||||||||||||||||||
Literature Links: |
LDHA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDHA - lactate dehydrogenase A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LDHC - lactate dehydrogenase C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002301.4 | 278 | Missense Mutation | TGT,TTT | C,F 35 | NP_002292.1 | |
NM_017448.3 | 278 | Missense Mutation | TGT,TTT | C,F 35 | NP_059144.1 | |
XM_017017721.1 | 278 | UTR 5 | XP_016873210.1 | |||
XM_017017722.1 | 278 | UTR 5 | XP_016873211.1 |