Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTATCAGAGAAGTCAGTAGAGG[C/T]CACTGAGACCGGCAGTCTTTCTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601387 | ||||||||||||||||||||
Literature Links: |
LDHAL6A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDHAL6A - lactate dehydrogenase A like 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144071.1 | 1303 | Intron | NP_001137543.1 | |||
NM_144972.4 | 1303 | Intron | NP_659409.2 | |||
XM_005252805.4 | 1303 | Intron | XP_005252862.2 | |||
XM_011519922.2 | 1303 | Intron | XP_011518224.1 | |||
XM_011519923.2 | 1303 | Intron | XP_011518225.1 | |||
XM_011519924.2 | 1303 | Intron | XP_011518226.1 | |||
XM_011519925.1 | 1303 | Intron | XP_011518227.1 |
TSG101 - tumor susceptibility 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006292.3 | 1303 | Missense Mutation | GAC,GGC | D,G 388 | NP_006283.1 | |
XM_005253108.3 | 1303 | Missense Mutation | GAC,GGC | D,G 336 | XP_005253165.1 |