Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGAAATCATTTCCACATCATCCA[C/T]CCACCCTCTTCTTAGGTAACAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602319 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NELL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NELL1 - neural EGFL like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288713.1 | Intron | NP_001275642.1 | ||||
NM_001288714.1 | Intron | NP_001275643.1 | ||||
NM_006157.4 | Intron | NP_006148.2 | ||||
NM_201551.2 | Intron | NP_963845.1 |