Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGAGAACTGGCGGAAAGACTT[C/T]GGATGAAAATGTCTGTTTCTCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604424 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIPK3 - homeodomain interacting protein kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001048200.2 | 309 | Intron | NP_001041665.1 | |||
NM_001278162.1 | 309 | UTR 5 | NP_001265091.1 | |||
NM_001278163.1 | 309 | Intron | NP_001265092.1 | |||
NM_005734.4 | 309 | Intron | NP_005725.3 | |||
XM_005252729.2 | 309 | UTR 5 | XP_005252786.1 | |||
XM_017017076.1 | 309 | Intron | XP_016872565.1 | |||
XM_017017077.1 | 309 | Intron | XP_016872566.1 |